Invitrogen
Manufacturer Code:PA557286
Catalog # PIPA557286
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human BXDC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brix domain containing 1; Brix domain-containing protein 1; homolog of Rpf2; ribosomal processing factor 2 homolog; Ribosome biogenesis protein RPF2 homolog; Ribosome production factor 2 homolog; RP11-397G5.1 BXDC1 bA397G5.4 brix domain containing 1 brix domain-containing protein 1 homolog of Rpf2 ribosomal processing factor 2 homolog ribosome biogenesis protein RPF2 homolog ribosome production factor 2 homolog
Gene Aliases: bA397G5.4; BXDC1; RPF2
UniProt ID: (Human) Q9H7B2
Entrez Gene ID: (Human) 84154
Molecular Function:
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.