Invitrogen
Manufacturer Code:PA542932
Catalog # PIPA542932
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human BVES |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Blood vessel epicardial substance; Blood vessel epicardial substance HBVES MGC42413 POP1 POPDC1; BVES; hBVES; mBVES; Pop1; popeye 1; popeye domain containing 1; popeye domain-containing 1; Popeye domain-containing protein 1; Popeye protein 1
Gene Aliases: BVES; HBVES; LGMD2X; mBVES; POP1; POPDC1; zgc:86887
UniProt ID: (Human) Q8NE79, (Rat) Q3BCU4, (Mouse) Q9ES83, (Zebrafish) Q5PQZ7
Entrez Gene ID: (Human) 11149, (Horse) 100066283, (Dog) 481944, (Rabbit) 100346465, (Guinea pig) 100726176, (Rat) 365603, (Mouse) 23828, (Zebrafish) 415107
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.