Invitrogen
Manufacturer Code:PA537134
Catalog # PIPA537134
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length human BVES |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Blood vessel epicardial substance; Blood vessel epicardial substance HBVES MGC42413 POP 1 POP1 POPDC 1 POPDC1 Popeye domain containing 1 Popeye domain containing protein 1 Popeye protein 1; hBVES; mBVES; popeye 1; popeye domain-containing 1; Popeye domain-containing protein 1; Popeye protein 1
Gene Aliases: BVES; HBVES; LGMD2X; mBVES; POP1; POPDC1
UniProt ID: (Human) Q8NE79, (Rat) Q3BCU4, (Mouse) Q9ES83
Entrez Gene ID: (Human) 11149, (Rat) 365603, (Mouse) 23828
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.