Novus Biologicals
Manufacturer Code:NBP299427AF350
Catalog # NB060574
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse BVES. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Blood vessel epicardial substance; blood vessel epicardial substance H HBVES POP1MGC42413 POPDC1popeye domain containing 1 Popeye domain-containing protein 1 Popeye protein 1; hBVES; Popeye domain-containing protein 1; Popeye protein 1
Gene Aliases: BVES; HBVES; LGMD2X; POP1; POPDC1
UniProt ID: (Human) Q8NE79
Entrez Gene ID: (Human) 11149
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.