Invitrogen
Manufacturer Code:PA5111014
Catalog # PIPA5111014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human BTRC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-transducin repeat-containing protein betaTrCP Beta-TrCP Beta-TrCP protein E3RS-IkappaB beta-TrCP1 BOS_23400 BTRC BTRCP b-TrCP bTrCP1 E3RSIkappaB E3RS-IkappaB Epididymis tissue protein Li 2a F-box and WD repeats protein beta-TrCP F-box/WD repeat-containing protein 1A Fbw1 FBW1A Fbxw1 FBXW1A Fwd1 HOS Kiaa4123 mE3RS-IkappaB pIkappaBalpha-E3 receptor subunit pIkappaB-E3 receptor subunit Slimb Ubiquitin ligase FWD1; beta-TrCP1; E3RSIkappaB; Epididymis tissue protein Li 2a; F-box and WD repeats protein beta-TrCP; F-box and WD-repeat protein 1B; F-box/WD repeat-containing protein 1A; pIkappaBalpha-E3 receptor subunit
Gene Aliases: BETA-TRCP; betaTrCP; BTRC; BTRCP; bTrCP1; FBW1A; FBXW1; FBXW1A; FWD1
UniProt ID: (Human) Q9Y297
Entrez Gene ID: (Human) 8945
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.