Invitrogen
Manufacturer Code:PA5109447
Catalog # PIPA5109447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 50-340 of human Beta-TRCP (NP_378663.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta-transducin repeat containing protein; Beta-transducin repeat-containing protein; Beta-transducin repeat-containing protein betaTrCP Beta-TrCP Beta-TrCP protein E3RS-IkappaB beta-TrCP1 BOS_23400 BTRC BTRCP b-TrCP bTrCP1 E3RSIkappaB E3RS-IkappaB Epididymis tissue protein Li 2a F-box and WD repeats protein beta-TrCP F-box/WD repeat-containing protein 1A Fbw1 FBW1A Fbxw1 FBXW1A Fwd1 HOS Kiaa4123 mE3RS-IkappaB pIkappaBalpha-E3 receptor subunit pIkappaB-E3 receptor subunit Slimb Ubiquitin ligase FWD1; Beta-TrCP; Beta-TrCP protein E3RS-IkappaB; beta-TrCP1; btrc; E3RSIkappaB; Epididymis tissue protein Li 2a; F-box and WD repeats protein beta-TrCP; F-box and WD-repeat protein 1B; F-box/WD repeat-containing protein 1A; HOS; mE3RS-IkappaB; pIkappaB-E3 receptor subunit; pIkappaBalpha-E3 receptor subunit; SCF b-TRCP; Ubiquitin ligase FWD1
Gene Aliases: b-TrCP; BETA-TRCP; Beta-Trcp1; betaTrCP; BTRC; BTRCP; bTrCP1; E3RS-IkappaB; E3RSIkappaB; Fbw1; FBW1A; FBXW1; FBXW1A; FWD1; HOS; Kiaa4123; Slimb
UniProt ID: (Human) Q9Y297, (Mouse) Q3ULA2
Entrez Gene ID: (Human) 8945, (Mouse) 12234, (Rat) 361765
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.