R&D Systems
Manufacturer Code:AF4917
Catalog # AF4917
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse BTN2A2/Butyrophilin 2A2 Gln29-Ser219 Accession # A4QPC6.1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: BTF2 BTF2BT2.2butyrophilin 2 BTN2 Butyrophilin 2 butyrophilin subfamily 2 member A2 butyrophilin subfamily 2 member A2 FLJ41908; butyrophilin 2; Butyrophilin subfamily 2 member A2; butyrophilin, subfamily 2, member A2
Gene Aliases: BT2.2; BTF2; BTN2.2; BTN2A2
UniProt ID: (Human) Q8WVV5
Entrez Gene ID: (Human) 10385
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.