Invitrogen
Manufacturer Code:MA531658
Catalog # PIMA531658
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4B11C8 |
Immunogen | Purified recombit fragment of human BTN2A2 (AA: extra 57-237) expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BTF2 BT2.2 BTN2.2; butyrophilin 2; Butyrophilin subfamily 2 member A2; butyrophilin, subfamily 2, member A2
Gene Aliases: BT2.2; BTF2; BTN2.2; BTN2A2
UniProt ID: (Human) Q8WVV5
Entrez Gene ID: (Human) 10385
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.