Invitrogen
Manufacturer Code:PA598743
Catalog # PIPA598743
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Butyrophilin subfamily 2 member A2 protein (33-262AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: butyrophilin 2; Butyrophilin subfamily 2 member A2; butyrophilin, subfamily 2, member A2
Gene Aliases: BT2.2; BTF2; Btn2; BTN2.2; BTN2A2; D030063K05
UniProt ID: (Human) Q8WVV5, (Mouse) A4QPC6
Entrez Gene ID: (Human) 10385, (Mouse) 238555
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.