Invitrogen
Manufacturer Code:PA547670
Catalog # PIPA547670
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse BTN2A2/Butyrophilin 2 Gln29-Ser219 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: BT2.2 BTF2 butyrophilin 2 butyrophilin subfamily 2 member A2; butyrophilin 2; Butyrophilin subfamily 2 member A2
Gene Aliases: Btn2; Btn2a2; D030063K05
UniProt ID: (Mouse) A4QPC6
Entrez Gene ID: (Mouse) 238555
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.