Novus Biologicals
Manufacturer Code:NB11087047
Catalog # NB11087047
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7F12H4 |
Immunogen | Purified recombinant C terminal fragment of BTK expressed in E. coli. [UniProt# Q06187] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: agammaglobulinaemia tyrosine kinase; Agammaglobulinaemia tyrosine kinase AGMX1MGC126262 AT ATKIMD1 B-cell progenitor kinase BPK Bruton agammaglobulinemia tyrosine kinase Bruton tyrosine kinase dominant-negative kinase-deficient Brutons tyrosine kinase EC 2.7.10 EC 2.7.10.2 PSCTK1 tyrosine-protein kinase BTK XLAMGC126261; Agammaglobulinemia tyrosine kinase; ATK; B-cell progenitor kinase; BPK; Bruton agammaglobulinemia tyrosine kinase; Bruton tyrosine kinase; Bruton's tyrosine kinase; dominant-negative kinase-deficient Brutons tyrosine kinase; truncated Bruton agammaglobulinemia tyrosine kinase; Tyrosine-protein kinase BTK; tyrosine-protein kinase BTK isoform (lacking exon 13 to 17); tyrosine-protein kinase BTK isoform (lacking exon 14)
Gene Aliases: AGMX1; AT; ATK; BPK; BTK; IMD1; PSCTK1; XLA
UniProt ID: (Human) Q06187
Entrez Gene ID: (Human) 695
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.