OriGene
Manufacturer Code:CF808151
Catalog # 50-166-8655
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2G5 |
Immunogen | Full length human recombit protein of human BTG2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: APRO1 PC3 TIS21; B-cell translocation gene 2; BTG family member 2; BTG family, member 2; nerve growth factor-inducible anti-proliferative; NGF-inducible anti-proliferative protein PC3; pheochromacytoma cell-3; Protein BTG2
Gene Aliases: APRO1; BTG2; PC3; TIS21
UniProt ID: (Human) P78543
Entrez Gene ID: (Human) 7832
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.