Invitrogen
Manufacturer Code:PA578888
Catalog # PIPA578888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence of human BTG2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-cell translocation gene 2; B-cell translocation gene 2, anti-proliferative; BTG family member 2; BTG family, member 2; cell surface alloantigen; Early induced gene B-cell translocation gene 2; nerve growth factor-inducible anti-proliferative; NGF-inducible anti-proliferative protein PC3; NGF-inducible anti-proliferative putative secreted protein; NGF-inducible protein TIS21; pheochromacytoma cell-3; Protein BTG2; Protein BTG2 BTG family member 2 NGF-inducible anti-proliferative protein PC3 BTG2 PC3
Gene Aliases: AA959598; Agl; An; an-1; APRO1; BTG2; PC3; TIS21
UniProt ID: (Human) P78543, (Rat) P27049, (Mouse) Q04211
Entrez Gene ID: (Human) 7832, (Rat) 29619, (Mouse) 12227
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.