Abnova
Manufacturer Code:H00007832A01
Catalog # 89-105-780
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 59 a.a. to 158 a.a) partial recombinant protein with GST tag.,BTG2 (NP_006754 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant BTG2.
For Research Use Only
Protein Aliases: B-cell translocation gene 2; BTG family member 2; BTG family, member 2; MGC126063; MGC126064; nerve growth factor-inducible anti-proliferative; NGF-inducible anti-proliferative protein PC3; PC3; pheochromacytoma cell-3; Protein BTG2; TIS21
Gene Aliases: APRO1; BTG2; PC3; TIS21
UniProt ID: (Human) P78543
Entrez Gene ID: (Human) 7832
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.