Proteintech
Manufacturer Code:163301AP150UL
Catalog # 50-173-0875
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BTD Fusion Protein Ag9449 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Biotinase; Biotinase biotinidase BTD; Biotinidase; btd {ECO:0000312|EMBL:AAH90017.1}
Gene Aliases: BTD
UniProt ID: (Human) P43251, (Rat) Q5FVF9, (Mouse) Q8CIF4
Entrez Gene ID: (Human) 686, (Rat) 306262, (Mouse) 26363
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.