Abnova
Manufacturer Code:H00114781B01
Catalog # 89-008-463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | BTBD9 (AAI01358.1 1 a.a. ~ 582 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human BTBD9 protein.
For Research Use Only
Protein Aliases: BTB (POZ) domain containing 9; BTB/POZ domain-containing protein 9; dJ322I12.1; FLJ32945; KIAA1880; MGC120517; MGC120519; MGC120520
Gene Aliases: BTBD9; dJ322I12.1; KIAA1880
UniProt ID: (Human) Q96Q07
Entrez Gene ID: (Human) 114781
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
hydrolase
non-motor actin binding protein
protease
serine protease
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.