Novus Biologicals
Manufacturer Code:NBP229622
Catalog # NBP229622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4F6 |
Immunogen | Full length recombinant human BST2 protein [NCBI NP_004326]. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Bone marrow stromal antigen 2; bone marrow stromal antigen 2 bone marrow stromal cell antigen 2 BST-2 CD317 CD317 antigen HM1.24 antigen NPC-A-7 PDCA-1 TETHERIN; BST-2; CD317; HM1.24 antigen; NPC-A-7; Tetherin
Gene Aliases: BST2; CD317; TETHERIN
UniProt ID: (Human) Q10589
Entrez Gene ID: (Human) 684
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.