OriGene
Manufacturer Code:CF501164
Catalog # 50-166-0198
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9H5 |
Immunogen | Full length human recombit protein of human BSG produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 5F7; 5F7 CD147 EMMPRIN OK TCSF; Basigin; basigin (Ok blood group); Basigin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); Basignin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); CD147; Collagenase stimulatory factor; EMMPRIN; Extracellular matrix metalloproteinase inducer; Glycoprotein CE9; HAb18G; Hepatoma-associated antigen; Leukocyte activation antigen M6; neurothelin; OK blood group antigen; OX-47 antigen; TCSF; Tumor cell-derived collagenase stimulatory factor
Gene Aliases: 5A11; 5F7; basignin; BSG; CD147; CE9; EMMPRIN; HT7; OK; Ox47R; TCSF; UNQ6505/PRO21383
UniProt ID: (Human) P35613, (Rat) P26453
Entrez Gene ID: (Human) 682, (Dog) 476758, (Rat) 25246
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.