Invitrogen
Manufacturer Code:PA531849
Catalog # PIPA531849
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 173 and 458 of Human BRUNOL4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bruno-like 4, RNA binding protein; Bruno-like protein 4; BRUNOL-4 BRUNOL4 Bruno -like 4 RNA binding protein CELF-4 CUG-BP and ETR-3 like factor 4 CUG-BP- and ETR-3-like factor 4 CUGBP Elav-like family member 4 LYST-interacting protein LIP9 RNA-binding protein BRUNOL-4 RNA-binding protein BRUNOL4 bruno-like 4 RNA binding prote; CELF-4; CUG-BP and ETR-3 like factor 4; CUG-BP- and ETR-3-like factor 4; CUGBP Elav-like family member 4; LYST-interacting protein LIP9; RNA-binding protein BRUNOL-4; RNA-binding protein BRUNOL4
Gene Aliases: BRUNOL4; CELF-4; CELF4
UniProt ID: (Human) Q9BZC1
Entrez Gene ID: (Human) 56853
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.