Invitrogen
Manufacturer Code:PA536436
Catalog # PIPA536436
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 20-70 of human BRP44L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis-regulating basic protein; Brain protein 44-like protein; HSPC040 protein; Mitochondrial pyruvate carrier 1; Mitochondrial pyruvate carrier 1 Brain protein 44-like proteinCGI-129 HSPC040 PNAS-115BRP44LMPC1
Gene Aliases: 0610006G08Rik; 3830411I18Rik; Arbp; BRP44L; CGI-129; dJ68L15.3; HSPC040; MPC1; MPYCD; PNAS-115
UniProt ID: (Human) Q9Y5U8, (Rat) P63031, (Mouse) P63030
Entrez Gene ID: (Human) 51660, (Rat) 171087, (Mouse) 55951
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.