Abnova
Manufacturer Code:PAB17635
Catalog # 89-119-298
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human BRP44L. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of BRP44L.
For Research Use Only
Protein Aliases: Brain protein 44-like protein; CGI-129; dJ68L15.3; HSPC040 protein; Mitochondrial pyruvate carrier 1
Gene Aliases: BRP44L; CGI-129; dJ68L15.3; HSPC040; MPC1; MPYCD; PNAS-115
UniProt ID: (Human) Q9Y5U8
Entrez Gene ID: (Human) 51660
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.