Novus Biologicals
Manufacturer Code:NBP280086
Catalog # NB013217
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | JCM-1 |
Immunogen | Peptide coupled to KLH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Brain protein 44; brain protein 44 DKFZP564B167 MGC125752 MGC125753; Mitochondrial pyruvate carrier 2
Gene Aliases: BRP44; MPC2
UniProt ID: (Human) O95563
Entrez Gene ID: (Human) 25874
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.