Invitrogen
Manufacturer Code:PA571855
Catalog # PIPA571855
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 98-127 amino acids from the C-terminal region of human BRP44 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain protein 44; Mitochondrial pyruvate carrier 2; Mitochondrial pyruvate carrier 2 Brain protein 44 MPC2 BRP44
Gene Aliases: BRP44; MPC2
UniProt ID: (Human) O95563
Entrez Gene ID: (Human) 25874
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.