Invitrogen
Manufacturer Code:PA5116939
Catalog # PIPA5116939
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human MPC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610006C01Rik 2010002I07Rik 2610205H19Rik AA108335 Brain protein 44 BRP44 ESTM43 Mitochondrial pyruvate carrier 2 MPC2 Protein 0-44 RGD1563422; Brain protein 44; Mitochondrial pyruvate carrier 2; Protein 0-44
Gene Aliases: 0610006C01Rik; 2010002I07Rik; 2610205H19Rik; AA108335; BRP44; ESTM43; MPC2; RGD1563422
UniProt ID: (Human) O95563, (Rat) P38718, (Mouse) Q9D023
Entrez Gene ID: (Human) 25874, (Rat) 100359982, (Mouse) 70456
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.