Proteintech
Manufacturer Code:119621AP150UL
Catalog # 50-172-7598
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MCPH/BRIT1 Fusion Protein Ag2567 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BRCT-repeat inhibitor of TERT expression 1; BRIT1 MCPH1 MCT Microcephalin microcephalin 1; Microcephalin; truncated microcephalin
Gene Aliases: 5430437K10Rik; BRIT1; D030046N04Rik; MCPH1; MCT
UniProt ID: (Human) Q8NEM0, (Mouse) Q7TT79
Entrez Gene ID: (Human) 79648, (Mouse) 244329
Molecular Function: DNA binding protein damaged DNA-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.