Invitrogen
Manufacturer Code:PA568973
Catalog # PIPA568973
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human BRF1/BRF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Butyrate response factor 1; Butyrate response factor 2; early response factor Berg36; EGF-inducible protein CMG1; EGF-response factor 1; EGF-response factor 1 butyrate response factor 1 early response factor Berg36 zinc finger protein 36 C3H1 type-like 1 zinc finger protein C3H type 36-like 1; EGF-response factor 2; ERF-1; ERF-2; mRNA decay activator protein ZFP36L1; mRNA decay activator protein ZFP36L2; TPA-induced sequence 11b; TPA-induced sequence 11d; ZFP36 ring finger protein-like 1; ZFP36 ring finger protein-like 2; ZFP36-like 1; ZFP36-like 2; zinc finger protein 36, C3H type-like 1; zinc finger protein 36, C3H type-like 2; zinc finger protein 36, C3H type-like 2-like; Zinc finger protein 36, C3H1 type-like 1; Zinc finger protein 36, C3H1 type-like 2; zinc finger protein, C3H type, 36-like 1; zinc finger protein, C3H type, 36-like 2
Gene Aliases: AW742437; AW743212; BERG36; BRF1; BRF2; Cmg1; D530020L18Rik; ERF-1; ERF-2; ERF1; ERF2; RNF162B; RNF162C; TIS11B; TIS11D; ZFP36L1; ZFP36L2
UniProt ID: (Human) Q07352, (Human) P47974, (Rat) P17431, (Mouse) P23950, (Mouse) P23949
Entrez Gene ID: (Human) 677, (Dog) 490748, (Rabbit) 100349183, (Bovine) 614773, (Horse) 100063597, (Bovine) 767898, (Human) 678, (Dog) 100856710, (Horse) 100069189, (Rabbit) 100351567, (Rat) 29344, (Mouse) 12192, (Guinea pig) 100721072, (Rat) 298765, (Guinea pig) 100724488, (Mouse) 12193
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.