Abnova
Manufacturer Code:H00000676B01P
Catalog # 89-032-119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | BRDT (AAI18499.1 1 a.a. ~ 947 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse Polyclonal Antibody
For Research Use Only
Protein Aliases: BRD6; Bromodomain testis-specific protein; bromodomain, testis-specific; Cancer/testis antigen 9; CT9; RING3-like protein
Gene Aliases: BRD6; BRDT; CT9
UniProt ID: (Human) Q58F21
Entrez Gene ID: (Human) 676
Molecular Function:
DNA binding protein
acetyltransferase
chromatin/chromatin-binding protein
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.