Invitrogen
Manufacturer Code:PA540818
Catalog # PIPA540818
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human BRDT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bromodomain testis-specific; Bromodomain testis-specific protein; bromodomain, testis-specific; Bromodomain-containing female sterile homeotic-like protein; Cancer/testis antigen 9; CT9; female sterile homeotic-related gene 3; RING3-like protein; testis-specific bromodomain protein
Gene Aliases: 7420412D09Rik; BRD6; BRDT; CT9; fi25h02; Fsrg3; si:dkeyp-85h7.3; wu:fi25h02
UniProt ID: (Human) Q58F21, (Mouse) Q91Y44
Entrez Gene ID: (Rabbit) 100355953, (Bovine) 523971, (Dog) 479954, (Horse) 100051647, (Human) 676, (Guinea pig) 100719653, (Mouse) 114642, (Rat) 305123, (Zebrafish) 333996
Molecular Function:
DNA binding protein
acetyltransferase
chromatin/chromatin-binding protein
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.