Invitrogen
Manufacturer Code:PA5117277
Catalog # PIPA5117277
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human BRDT. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BRD6 BRDT bromodomain testis associated bromodomain testis-specific protein bromodomain testis-specific cancer/testis antigen 9 CT9 RING3-like protein; Bromodomain testis-specific protein; Bromodomain-containing female sterile homeotic-like protein; female sterile homeotic-related gene 3; RING3-like protein
Gene Aliases: 7420412D09Rik; Brd6; Brdt; Fsrg3
UniProt ID: (Mouse) Q91Y44
Entrez Gene ID: (Mouse) 114642
Molecular Function: DNA binding protein acetyltransferase chromatin/chromatin-binding protein nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.