Invitrogen
Manufacturer Code:PA530413
Catalog # PIPA530413
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 641 and 876 of Human BRD8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bromodomain-containing protein 8; p120; Skeletal muscle abundant protein; Skeletal muscle abundant protein 2; SMAP SMAP2 p120 bromodomain-containing protein 8 skeletal muscle abundant protein 2 thyroid hormone receptor coactivating protein of 120 kDa trCP120; Thyroid hormone receptor coactivating protein of 120 kDa; TrCP120
Gene Aliases: BRD8; p120; SMAP; SMAP2
UniProt ID: (Human) Q9H0E9
Entrez Gene ID: (Human) 10902
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.