Novus Biologicals
Manufacturer Code:NB10079780
Catalog # NB10079780
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 25 and 75 of human Bromodomain containing 8 using the numbering given in TrEMBL entry Q969M9 (GeneID 10902). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bromodomain containing 8 bromodomain-containing protein 8 Skeletal muscle abundant protein SMAP2 SMAPp120Skeletal muscle abundant protein 2 Thyroid hormone receptor coactivating protein of 120 kDa trCP120; Bromodomain-containing protein 8; p120; Skeletal muscle abundant protein; Skeletal muscle abundant protein 2; Thyroid hormone receptor coactivating protein of 120 kDa; TrCP120
Gene Aliases: BRD8; p120; SMAP; SMAP2
UniProt ID: (Human) Q9H0E9
Entrez Gene ID: (Human) 10902
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.