Proteintech
Manufacturer Code:510092AP150UL
Catalog # 50-560-231
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BRD7 Fusion Protein Ag3911 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 75 kDa bromodomain protein; 75 kDa bromodomain protein BP75 BRD7 bromodomain containing 7 CELTIX1 NAG4 Protein CELTIX 1; bromodomain protein 75 kDa; bromodomain-containing 7; Bromodomain-containing protein 7; Protein CELTIX-1; protein tyrosine phosphatase, non-receptor type 13 interacting protein
Gene Aliases: BP75; BRD7; CELTIX1; NAG4; Ptpn13ip
UniProt ID: (Human) Q9NPI1, (Mouse) O88665
Entrez Gene ID: (Human) 29117, (Mouse) 26992, (Rat) 361374
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.