Novus Biologicals
Manufacturer Code:NBP276394
Catalog # NBP276394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | BL-151-6F11 |
Immunogen | Immunogen sequence between amino acids 1312 and 1362 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 2-8°C/ 1 year from date of receipt |
Rabbit Monoclonal Antibody
Protein Aliases: bromodomain containing 4 bromodomain-containing 4 CAPchromosome-associated protein HUNK1bromodomain-containing protein 4 HUNKI MCAP Protein HUNK1; bromodomain-containing 4; Bromodomain-containing protein 4; chromosome-associated protein; Protein HUNK1
Gene Aliases: BRD4; CAP; HUNK1; HUNKI; MCAP
UniProt ID: (Human) O60885
Entrez Gene ID: (Human) 23476
Molecular Function: DNA binding protein acetyltransferase chromatin/chromatin-binding protein nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.