Invitrogen
Manufacturer Code:PA141403
Catalog # 01-675-532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 150-200 of human BRD4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bromodomain-containing 4; bromodomain-containing 5; Bromodomain-containing protein 4; chromosome-associated protein; fsh-like protein; HUNK1; MCAP; Mitotic chromosome-associated protein; Protein HUNK1
Gene Aliases: BRD4; Brd5; CAP; HUNK1; HUNKI; MCAP; WI-11513
UniProt ID: (Human) O60885, (Mouse) Q9ESU6
Entrez Gene ID: (Human) 23476, (Rat) 362844, (Mouse) 57261
Molecular Function: DNA binding protein acetyltransferase chromatin/chromatin-binding protein nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.