Novus Biologicals
Manufacturer Code:NBP275422
Catalog # NBP275422
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG40-42 |
Immunogen | Recombinant protein within human BRD2 aa 1-200. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: bromodomain containing 2 bromodomain-containing 2 bromodomain-containing protein 2 D6S113E female sterile homeotic-related gene 1 FSRG1FSH KIAA9001FLJ31942 NAT O27.1.1 Really interesting new gene 3 protein RING3DKFZp686N0336 RNF3; Bromodomain-containing protein 2; female sterile homeotic-related gene 1; O27.1.1; Really interesting new gene 3 protein
Gene Aliases: BRD2; D6S113E; FSH; FSRG1; KIAA9001; NAT; O27.1.1; RING3; RNF3
UniProt ID: (Human) P25440
Entrez Gene ID: (Human) 6046
Molecular Function:
DNA binding protein
acetyltransferase
chromatin/chromatin-binding protein
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.