Invitrogen
Manufacturer Code:PA583631
Catalog # PIPA583631
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human BRD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bromodomain-containing protein 2; D6S113E FSRG1 KIAA9001 NAT RING3; female sterile homeotic-related gene 1; O27.1.1; Really interesting new gene 3 protein
Gene Aliases: BRD2; D6S113E; FSH; FSRG1; KIAA9001; NAT; O27.1.1; RING3; RNF3
UniProt ID: (Human) P25440
Entrez Gene ID: (Human) 6046
Molecular Function:
DNA binding protein
acetyltransferase
chromatin/chromatin-binding protein
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.