Invitrogen
Manufacturer Code:PA5102953
Catalog # PIPA5102953
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BRCC3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6.1A protein; 6.1A protein BRCA1/BRCA2-containing complex subunit 3 BRCA1/BRCA2-containing complex subunit 36 BRCA1/BRCA2-containing complex subunit 3 BRCA1-A complex subunit BRCC36 Brcc3 Brcc36 BRISC complex subunit BRCC36 C6.1A CXorf53 Lys-63-specific deubiquitinase BRCC36 RP11-143H17.2; BRCA1-A complex subunit BRCC36; BRCA1/BRCA2-containing complex subunit 3; BRCA1/BRCA2-containing complex subunit 36; BRISC complex subunit BRCC36; Lys-63-specific deubiquitinase BRCC36
Gene Aliases: BRCC3; BRCC36; C6.1A; CXorf53
UniProt ID: (Human) P46736, (Rat) B2RYM5, (Mouse) P46737
Entrez Gene ID: (Human) 79184, (Rat) 316794, (Mouse) 210766
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.