Novus Biologicals
Manufacturer Code:NBP141187
Catalog # NBP141187
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 9D3 |
Immunogen | BRCA2 protein fragment containing amino acids 2336-2478 of human BRCA2 expressed in E. coli. BRCA2-P400. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: BRCA1/BRCA2-containing complex subunit 2 BRCC2 breast and ovarian cancer susceptibility gene early onset breast cancer 2 tumor suppressor breast cancer 2 early onset breast cancer susceptibility protein BRCA2 breast cancer type 2 susceptibility protein BROVCA2 FACD FAD FAD1 FANCB FANCD FANCD1Fanconi anemia complementation group D1 Fanconi anemia group D1 protein GLM3 PNCA2; BRCA1/BRCA2-containing complex, subunit 2; breast and ovarian cancer susceptibility gene, early onset; breast and ovarian cancer susceptibility protein 2; breast cancer 2 tumor suppressor; breast cancer 2, early onset; Breast cancer type 2 susceptibility protein; Fanconi anemia group D1 protein; mutant BRCA2
Gene Aliases: BRCA2; BRCC2; BROVCA2; FACD; FAD; FAD1; FANCD; FANCD1; GLM3; PNCA2; XRCC11
UniProt ID: (Human) P51587
Entrez Gene ID: (Human) 675
Molecular Function: DNA binding protein damaged DNA-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.