Invitrogen
Manufacturer Code:PA5105731
Catalog # PIPA5105731
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BRCA2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brca 2 BRCA1/BRCA2-containing complex subunit 2 Brca2 brca-2 BRCA2 DNA repair associated BRCC2 breast and ovarian cancer susceptibility gene early onset breast and ovarian cancer susceptibility protein 2 breast cancer 2 breast cancer 2 tumor suppressor breast cancer 2 early onset breast cancer 2 mutation 1 University of Wisconsin-Madison breast cancer susceptibility protein 2 breast cancer type 2 susceptibility protein Breast cancer type 2 susceptibility protein homolog BROVCA2 FACD FAD FAD1 FANCB FANCD FANCD1 Fanconi anemia group D1 protein fanconi anemia group D1 protein homolog GLM3 mutant BRCA2 PNCA2 RAB163 RP11-298P3.4 XRCC11; BRCA1/BRCA2-containing complex, subunit 2; breast and ovarian cancer susceptibility gene, early onset; breast and ovarian cancer susceptibility protein 2; breast cancer 2 tumor suppressor; breast cancer 2, early onset; breast cancer 2, mutation 1, University of Wisconsin-Madison; breast cancer susceptibility protein 2; Breast cancer type 2 susceptibility protein; Breast cancer type 2 susceptibility protein homolog; Fanconi anemia group D1 protein; Fanconi anemia group D1 protein homolog; mutant BRCA2
Gene Aliases: BRCA2; BRCC2; BROVCA2; FACD; FAD; FAD1; FANCD; FANCD1; GLM3; PNCA2; RAB163; XRCC11
UniProt ID: (Human) P51587, (Mouse) P97929
Entrez Gene ID: (Human) 675, (Mouse) 12190, (Rat) 360254
Molecular Function:
DNA binding protein
damaged DNA-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.