Novus Biologicals
Manufacturer Code:NBP237435
Catalog # NBP237435
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F11E3 |
Immunogen | Purified recombinant fragment of mouse Lplunc1 (AA: 248-475) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bA49G10.6 chromosome 20 open reading frame 114 dJ1187J4.1 long palate lung and nasal epithelium carcinoma associated 1 long palate lung and nasal epithelium carcinoma-associated protein 1 LPLUNC1 MGC14597 VEMSGP Von Ebner minor salivary gland protein; BPI fold containing family B, member 1; BPI fold-containing family B member 1; long palate, lung and nasal epithelium carcinoma associated 1; Long palate, lung and nasal epithelium carcinoma-associated protein 1; VEMSGP; von Ebner minor salivary gland protein
Gene Aliases: BPIFB1; C20orf114; LPLUNC1; UNQ706/PRO1357
UniProt ID: (Human) Q8TDL5
Entrez Gene ID: (Human) 92747
Molecular Function: antibacterial response protein defense/immunity protein transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.