R&D Systems
Manufacturer Code:IC74681G100UG
Catalog # IC74681G10
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 971526 |
Immunogen | Human embryonic kidney cell line HEK-293-derived transfected with human BPI Val32-Lys487 Accession # P17213 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: Bactericidal permeability-increasing protein; bactericidal permeability-increasing protein bactericidal/permeability-increasing protein BPIFD1 CAP 57 rBPI; BPI; BPI fold containing family D, member 1; CAP 57; recombinant BPI holoprotein, rBPI
Gene Aliases: BPI; BPIFD1; rBPI
UniProt ID: (Human) P17213
Entrez Gene ID: (Human) 671
Molecular Function:
antibacterial response protein
defense/immunity protein
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.