Invitrogen
Manufacturer Code:MA530095
Catalog # PIMA530095
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 11 |
Immunogen | Recombit Human BPI Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Bactericidal permeability-increasing protein; BPI; BPI fold containing family D, member 1; CAP 57; recombinant BPI holoprotein, rBPI; RP13-218F6&lowbarA.1
Gene Aliases: BPI; BPIFD1; rBPI
UniProt ID: (Human) P17213
Entrez Gene ID: (Human) 671
Molecular Function:
antibacterial response protein
defense/immunity protein
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.