Invitrogen
Manufacturer Code:PA5102736
Catalog # PIPA5102736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BPI corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9230105K17Rik Bactericidal permeability-increasing protein bactericidal permeablility increasing protein bactericidal/permeability-increasing protein BPI BPI fold containing family D member 1 BPIFD1 CAP 57 rBPI recombinant BPI holoprotein rBPI RP13-218F6_A.1; Bactericidal permeability-increasing protein; BPI; BPI fold containing family D, member 1; CAP 57; recombinant BPI holoprotein, rBPI
Gene Aliases: BPI; BPIFD1; rBPI
UniProt ID: (Human) P17213, (Rat) Q6AXU0
Entrez Gene ID: (Human) 671, (Rat) 296321
Molecular Function:
antibacterial response protein
defense/immunity protein
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.