Invitrogen
Manufacturer Code:PA557798
Catalog # PIPA557798
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human BPHL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Biphenyl hydrolase-like protein; Biphenyl hydrolase-related protein; Bph-rp; Breast epithelial mucin-associated antigen; MCNAA; Poly(ADP-ribose) glycohydrolase; VACVase; Valacyclovir hydrolase; Valacyclovir hydrolase VACVase Valacyclovirase 31-- Biphenyl hydrolase-like protein Biphenyl hydrolase-related protein Bph-rp Breast epithelial mucin-associated antigen MCNAA BPHL MCNAA; valacyclovirase
Gene Aliases: BPH-RP; BPHL; MCNAA; PARG; VACVASE
UniProt ID: (Human) Q86WA6
Entrez Gene ID: (Human) 670
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.