Novus Biologicals
Manufacturer Code:NBP250124AF532
Catalog # NP20124A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | T7-P1B7&asteriskA8 |
Immunogen | Peptide Sequence - KGHVLTRDLG |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: block of proliferation 1 KIAA0124Block of proliferation 1 protein ribosome biogenesis protein BOP1; Block of proliferation 1 protein; Ribosome biogenesis protein BOP1
Gene Aliases: BOP1; KIAA0124
UniProt ID: (Human) Q969Z6
Entrez Gene ID: (Human) 23246
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.