Novus Biologicals
Manufacturer Code:NBP147308
Catalog # NBP147308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 169 and 219 of human BCL2/Adenovirus E1B 19kDa Interacting Protein 3-Like using the numbering given in entry NP_004322.1 (GeneID 665). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
For Research Use Only
Protein Aliases: Adenovirus E1B19K-binding protein B5; Adenovirus E1B19K-binding protein B5 BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like BCL2/adenovirus E1B 19-kd protein-interacting protein 3a BCL2/adenovirus E1B 19kDa interacting protein 3-like BNIP3a BNIP3H NIP3L NIP3-like protein X Nix NIXBCL2/adenovirus E1B 19kD-interacting protein 3-like; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A; BCL2/adenovirus E1B 19-kd protein-interacting protein 3a; BCL2/adenovirus E1B 19kDa interacting protein 3 like; BCL2/adenovirus E1B 19kDa interacting protein 3-like; NIP3-like protein X; NIP3L
Gene Aliases: BNIP3A; BNIP3H; BNIP3L; NIX
UniProt ID: (Human) O60238
Entrez Gene ID: (Human) 665
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.