Novus Biologicals
Manufacturer Code:NB11060562SS
Catalog # NB11060562T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide against an N-terminal region (within residues 1-50) of the human BNIP3 protein. [UniProt# Q12983] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BCL2/adenovirus E1B 19 kDa protein-interacting protein 3; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3 BCL2/adenovirus E1B 19kDa interacting protein 3 BCL2/adenovirus E1B 19kD-interacting protein 3 NIP3; BCL2/adenovirus E1B 19kDa interacting protein 3
Gene Aliases: BNIP3; NIP3
UniProt ID: (Human) Q12983
Entrez Gene ID: (Human) 664
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.