OriGene
Manufacturer Code:CF802188
Catalog # 50-166-5893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-199 of human BNIP1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BCL2/adenovirus E1B 19 kDa protein-interacting protein 1; BCL2/adenovirus E1B 19kDa interacting protein 1; NIP1 SEC20 TRG-8; Transformation-related gene 8 protein; TRG-8; Vesicle transport protein SEC20
Gene Aliases: BNIP1; NIP1; SEC20; SEC20L; TRG-8; TRG8
UniProt ID: (Human) Q12981
Entrez Gene ID: (Human) 662
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.