Invitrogen
Manufacturer Code:PA544993
Catalog # PIPA544993
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human BNIP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BCL2/adenovirus E1B 19 kDa protein-interacting protein 1; BCL2/adenovirus E1B 19kD interacting protein 1; BCL2/adenovirus E1B 19kDa interacting protein 1; BCL2/adenovirus E1B 19kDa interacting protein 1 NIP1 SEC20 TRG-8; BCL2/adenovirus E1B 19kDa-interacting protein 1; BCL2/adenovirus E1B 19kDa-interacting protein 1, NIP1; BCL2/adenovirus E1B interacting protein 1, NIP1; BCL2/adenovirus E1B interacting protein 1-like 1; bnip1; bnip1 {ECO:0000312|EMBL:AAL50991.1, ECO:0000312|RGD:620799}; SNARE protein SEC20; Transformation-related gene 8 protein; TRG-8; Vesicle transport protein SEC20
Gene Aliases: 2010005M06Rik; 5930429G21Rik; BNIP1; Bnip1l1; NIP1; SEC20; SEC20L; TRG-8; TRG8
UniProt ID: (Human) Q12981, (Mouse) Q6QD59, (Rat) Q8VHI8
Entrez Gene ID: (Human) 662, (Horse) 100069728, (Bovine) 614520, (Rabbit) 100342023, (Dog) 609594, (Mouse) 224630, (Rat) 140932, (Guinea pig) 100726766
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.