Invitrogen
Manufacturer Code:PA537215
Catalog # PIPA537215
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in human BMS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BMS1 homolog, ribosome assembly protein; BMS1 ribosome biogenesis factor; BMS1-like, ribosome assembly protein; Ribosome assembly protein BMS1 homolog; Ribosome biogenesis protein BMS1 homolog; Ribosome biogenesis protein BMS1 homolog Ribosome assembly protein BMS1 homolog BMS1 BMS1L KIAA0187
Gene Aliases: AA408648; ACC; AU020092; AU043373; BB007109; BC030906; BMS1; BMS1L; KIAA0187; mKIAA0187
UniProt ID: (Human) Q14692
Entrez Gene ID: (Human) 9790, (Mouse) 213895, (Rat) 362426
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.